ENA429359511 Enamine

2-bromo-2-(3,4-dichlorophenyl)acetic acid

  • MDL number MFCD23701419
  • Cas Number : 72191-59-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-188023

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-188023
Availability details for
Quantity
  • Availability
  • Add to Cart