ENA429359514 Enamine

2-(aminomethyl)-5-fluorophenol hydrobromide

  • MDL number MFCD28118683
  • Cas Number : 1803588-61-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HBr
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-188032

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-188032
Availability details for
Quantity
  • Availability
  • Add to Cart