ENA429359515 Enamine

2-(aminomethyl)-5-fluorophenol

  • MDL number MFCD16293420
  • Cas Number : 870669-90-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-188033

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-188033
Availability details for
Quantity
  • Availability
  • Add to Cart