ENA429359519 Enamine

methyl 4-aminopiperidine-1-carboxylate hydrochloride

  • MDL number MFCD18377888
  • Cas Number : 1187160-88-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-188041

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-188041
Availability details for
Quantity
  • Availability
  • Add to Cart