ENA429359524 Enamine

imidazo[1,2-a]pyridin-7-amine dihydrochloride

  • MDL number MFCD24368618
  • Cas Number : 1427195-25-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-188049

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-188049
Availability details for
Quantity
  • Availability
  • Add to Cart