ENA429359527 Enamine

4-bromo-5-oxo-1-phenyl-2,5-dihydro-1H-pyrazole-3-carboxylic acid

  • MDL number MFCD28118686
  • Cas Number : 1803572-42-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-188064

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-188064
Availability details for
Quantity
  • Availability
  • Add to Cart