ENA429359528 Enamine

methyl 5-bromo-3-cyano-6-methyl-2-oxo-1,2-dihydropyridine-4-carboxylate

  • MDL number MFCD28118687
  • Cas Number : 1630763-53-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-188066

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-188066
Availability details for
Quantity
  • Availability
  • Add to Cart