ENA429359530 Enamine

1-amino-5-bromo-4,6-dimethyl-2-oxo-1,2-dihydropyridine-3-carbonitrile

  • MDL number MFCD28097874
  • Cas Number : 1630763-65-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-188069

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-188069
Availability details for
Quantity
  • Availability
  • Add to Cart