ENA429359534 Enamine

tert-butyl 5-bromo-2,4-dioxo-1,2,3,4-tetrahydropyrimidine-1-carboxylate

  • MDL number MFCD24475089
  • Cas Number : 402848-99-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-188086

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-188086
Availability details for
Quantity
  • Availability
  • Add to Cart