ENA429359537 Enamine

2-(N-methylprop-2-enamido)acetic acid

  • MDL number MFCD09937931
  • Cas Number : 71325-63-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-188089

Safety Information

msds https://www.enaminestore.com/catalog/EN300-188089
Availability details for
Quantity
  • Availability
  • Add to Cart