ENA429359538 Enamine

2-phenoxybutanoyl chloride

  • MDL number MFCD00051569
  • Cas Number : 41717-60-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-188091

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-188091
Availability details for
Quantity
  • Availability
  • Add to Cart