ENA429359539 Enamine

oxepane-4-carboxylic acid

  • MDL number MFCD21124931
  • Cas Number : 933747-23-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-188092

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-188092
Availability details for
Quantity
  • Availability
  • Add to Cart