ENA429359540 Enamine

4-(diethylamino)benzene-1-carbothioamide

  • MDL number MFCD11135384
  • Cas Number : 772324-97-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-188094

Safety Information

msds https://www.enaminestore.com/catalog/EN300-188094
Availability details for
Quantity
  • Availability
  • Add to Cart