ENA429359541 Enamine

4-phenoxypyrimidin-2-amine

  • MDL number MFCD00463193
  • Cas Number : 882767-05-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-188095

Safety Information

msds https://www.enaminestore.com/catalog/EN300-188095
Availability details for
Quantity
  • Availability
  • Add to Cart