ENA429359546 Enamine

1-phenylpiperidine-2,4-dione

  • MDL number MFCD21944048
  • Cas Number : 99539-86-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. -20
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-188102

Safety Information

msds https://www.enaminestore.com/catalog/EN300-188102
Availability details for
Quantity
  • Availability
  • Add to Cart