ENA429359548 Enamine

4-(aminomethyl)-N-hydroxybenzamide hydrochloride

  • MDL number MFCD28118689
  • Cas Number : 1803600-37-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-188104

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-188104
Availability details for
Quantity
  • Availability
  • Add to Cart