ENA429359552 Enamine

2-[(2-sulfanylethyl)sulfanyl]ethan-1-ol

  • MDL number MFCD28118691
  • Cas Number : 98026-19-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-188108

Safety Information

signalword Danger
pcodes P210,P233,P240,P241,P242,P243,P264,P270,P280,P301+P312,P302+P352,P303+P361+P353,P305+P351+P338,P312,P330,P332+P313,P337+P313,P361,P362,P363,P370+P378,P403+P235,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-188108
pictograms GHS02,GHS06
hstatements H226,H302,H311,H315,H319
Availability details for
Quantity
  • Availability
  • Add to Cart