ENA429359556 Enamine

6-chloro-2-ethoxypyridine-3-carboxylic acid

  • MDL number MFCD18343601
  • Cas Number : 1343080-22-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-188112

Safety Information

msds https://www.enaminestore.com/catalog/EN300-188112
Availability details for
Quantity
  • Availability
  • Add to Cart