ENA429359558 Enamine

[2-(4-fluorophenyl)-1H-imidazol-5-yl]methanol

  • MDL number MFCD10001459
  • Cas Number : 191158-31-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-188114

Safety Information

msds https://www.enaminestore.com/catalog/EN300-188114
Availability details for
Quantity
  • Availability
  • Add to Cart