ENA429359560 Enamine

5-bromo-2-methyl-4-[(4-methyl-1,2,5-oxadiazol-3-yl)methoxy]pyrimidine

  • MDL number MFCD28118692
  • Cas Number : 1803598-74-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-188118

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-188118
Availability details for
Quantity
  • Availability
  • Add to Cart