ENA429359562 Enamine

N-(2-methylphenyl)-2-(propylamino)propanamide

  • MDL number MFCD00048681
  • Cas Number : 721-50-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-188120

Safety Information

msds https://www.enaminestore.com/catalog/EN300-188120
Availability details for
Quantity
  • Availability
  • Add to Cart