ENA429359566 Enamine

4-ethynylcyclohexan-1-one

  • MDL number MFCD26030888
  • Cas Number : 1549050-50-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. -20
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-188124

Safety Information

signalword Danger
msds https://www.enaminestore.com/catalog/EN300-188124
pictograms GHS02
Availability details for
Quantity
  • Availability
  • Add to Cart