ENA429359576 Enamine

4-[(2-oxo-1,2,3,4-tetrahydroquinolin-7-yl)oxy]butanoic acid

  • MDL number MFCD28118698
  • Cas Number : 58899-27-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-188172

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-188172
Availability details for
Quantity
  • Availability
  • Add to Cart