ENA429359578 Enamine

2-oxo-2H,3H-[1,3]oxazolo[4,5-b]pyridine-5-carboxylic acid

  • MDL number MFCD20647777
  • Cas Number : 61548-55-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-188174

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-188174
Availability details for
Quantity
  • Availability
  • Add to Cart