ENA429359581 Enamine

3-fluoro-2-(1H-1,2,4-triazol-3-yl)aniline

  • MDL number MFCD14640949
  • Cas Number : 1249534-87-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-188181

Safety Information

msds https://www.enaminestore.com/catalog/EN300-188181
Availability details for
Quantity
  • Availability
  • Add to Cart