ENA429359584 Enamine

potassium 3-methyl-1,2,4-oxadiazole-5-carboxylate

  • MDL number MFCD16990593
  • Cas Number : 1240605-84-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data K+
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-188184

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-188184
Availability details for
Quantity
  • Availability
  • Add to Cart