ENA429359586 Enamine

6-(1-methyl-1H-pyrazol-4-yl)-2,3-dihydropyridazin-3-one

  • MDL number MFCD16109164
  • Cas Number : 1100598-49-9
  • Molecular Weight : 176.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-188186

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-188186
Availability details for
Quantity
  • Availability
  • Add to Cart