ENA429359590 Enamine

[1-(methylsulfanyl)cyclobutyl]methanamine

  • MDL number MFCD21127196
  • Cas Number : 1487438-51-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-188197

Safety Information

msds https://www.enaminestore.com/catalog/EN300-188197
Availability details for
Quantity
  • Availability
  • Add to Cart