ENA429359591 Enamine

2-amino-3-nitropyridine-4-carboxylic acid

  • MDL number MFCD28118700
  • Cas Number : 1795298-56-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-188209

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-188209
Availability details for
Quantity
  • Availability
  • Add to Cart