ENA429359597 Enamine

5-(5-fluoropyridin-3-yl)pyrimidin-2-amine

  • MDL number MFCD24266902
  • Cas Number : 1551820-69-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-188223

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-188223
Availability details for
Quantity
  • Availability
  • Add to Cart