ENA429359598 Enamine

3-acetyl-4-methylbenzoic acid

  • MDL number MFCD14536161
  • Cas Number : 1216050-56-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-188224

Safety Information

msds https://www.enaminestore.com/catalog/EN300-188224
Availability details for
Quantity
  • Availability
  • Add to Cart