ENA429359600 Enamine

5-amino-1-methyl-2,3-dihydro-1H-indol-2-one hydrochloride

  • MDL number MFCD28118701
  • Cas Number : 1790361-51-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-188228

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-188228
Availability details for
Quantity
  • Availability
  • Add to Cart