ENA429359602 Enamine

4-chloro-2-(3-chlorophenyl)aniline

  • MDL number MFCD24078223
  • Cas Number : 1521784-05-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-188230

Safety Information

msds https://www.enaminestore.com/catalog/EN300-188230
Availability details for
Quantity
  • Availability
  • Add to Cart