ENA429359608 Enamine

6-(2-chloroacetyl)-3-methyl-1,2,3,4-tetrahydroquinazolin-2-one

  • MDL number MFCD23715562
  • Cas Number : 118621-05-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-188240

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-188240
Availability details for
Quantity
  • Availability
  • Add to Cart