ENA429359609 Enamine

sodium 2-hydroxy-2-(pyridin-2-yl)acetate

  • MDL number MFCD28118704
  • Cas Number : 1706465-12-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data Na+
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-188242

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-188242
Availability details for
Quantity
  • Availability
  • Add to Cart