ENA429359614 Enamine

1-(2,5-difluorophenyl)-2,2-difluoroethan-1-amine

  • MDL number MFCD16618193
  • Cas Number : 1038724-69-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-188248

Safety Information

msds https://www.enaminestore.com/catalog/EN300-188248
Availability details for
Quantity
  • Availability
  • Add to Cart