ENA429359615 Enamine

1-iodo-2,4-bis(propan-2-yl)benzene

  • MDL number MFCD00052396
  • Cas Number : 496801-02-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-188250

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-188250
Availability details for
Quantity
  • Availability
  • Add to Cart