ENA429359617 Enamine

[(4-chlorophenyl)(cyclopropyl)methyl](methyl)amine

  • MDL number MFCD12179544
  • Cas Number : 1153551-29-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-188252

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-188252
Availability details for
Quantity
  • Availability
  • Add to Cart