ENA429359619 Enamine

1-hydroxy-1,3-dihydro-2,1-benzoxaborole-5-carbonitrile

  • MDL number MFCD22393302
  • Cas Number : 905710-80-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-188256

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-188256
Availability details for
Quantity
  • Availability
  • Add to Cart