ENA429359620 Enamine

5-amino-2-methoxy-N-(2-methoxyethyl)benzene-1-sulfonamide

  • MDL number MFCD11118556
  • Cas Number : 1036596-08-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-188258

Safety Information

msds https://www.enaminestore.com/catalog/EN300-188258
Availability details for
Quantity
  • Availability
  • Add to Cart