ENA429359621 Enamine

2-chloro-N-(4-methylbenzenesulfonyl)acetamide

  • MDL number MFCD00723359
  • Cas Number : 14949-06-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-188259

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-188259
Availability details for
Quantity
  • Availability
  • Add to Cart