ENA429359622 Enamine

4-acetyl-3-chlorobenzonitrile

  • MDL number MFCD26806424
  • Cas Number : 1096666-21-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-188260

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-188260
Availability details for
Quantity
  • Availability
  • Add to Cart