ENA429359623 Enamine

4-chloro-2-(naphthalen-1-yl)aniline

  • MDL number MFCD23545431
  • Cas Number : 1429791-79-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-188261

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-188261
Availability details for
Quantity
  • Availability
  • Add to Cart