ENA429359624 Enamine

2-fluoro-4,6-dimethylaniline

  • MDL number MFCD26938999
  • Cas Number : 1780776-35-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-188262

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-188262
Availability details for
Quantity
  • Availability
  • Add to Cart