ENA429359628 Enamine

2-[2-(2-nitrophenyl)ethyl]benzoic acid

  • MDL number MFCD28118709
  • Cas Number : 1803593-67-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-188268

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-188268
Availability details for
Quantity
  • Availability
  • Add to Cart