ENA429359630 Enamine

5-(difluoromethyl)-1-(pyridin-4-yl)-1H-pyrazol-4-amine

  • MDL number MFCD21773908
  • Cas Number : 1504808-16-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-188270

Safety Information

msds https://www.enaminestore.com/catalog/EN300-188270
Availability details for
Quantity
  • Availability
  • Add to Cart