ENA429359631 Enamine

1-(2-chlorophenyl)-3,3,3-trifluoropropan-1-amine

  • MDL number MFCD14657033
  • Cas Number : 1250845-35-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-188271

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-188271
Availability details for
Quantity
  • Availability
  • Add to Cart