ENA429359632 Enamine

O-[(4-methylmorpholin-2-yl)methyl]hydroxylamine

  • MDL number MFCD21920874
  • Cas Number : 1540203-63-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-188272

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-188272
Availability details for
Quantity
  • Availability
  • Add to Cart