ENA429359639 Enamine

5-amino-2-methoxy-N-(oxolan-3-yl)benzene-1-sulfonamide

  • MDL number MFCD23464280
  • Cas Number : 1540739-79-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-188290

Safety Information

msds https://www.enaminestore.com/catalog/EN300-188290
Availability details for
Quantity
  • Availability
  • Add to Cart