ENA429359641 Enamine

3,3-dimethyl-2,3-dihydro-1-benzofuran

  • MDL number MFCD17015650
  • Cas Number : 13524-78-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-188296

Safety Information

msds https://www.enaminestore.com/catalog/EN300-188296
Availability details for
Quantity
  • Availability
  • Add to Cart